129775-100G
129775-25G
129775-5G
BT129775-100GEA
275
USD
InStock
BT129775-100G
BT129775-25G
BT129775-5G
Pentamethylbenzene ≥99%
Pentamethylbenzene
CAS: 700-12-9; EC No: 211-837-8; MDL No: MFCD00008522
UN No: UN1325; Haz Class: 4.1; Packing Group: III
Crystalline; Linear Formula: C6H(CH3)5; Molecular Formula: C11H16; MW: 148.25
Melting Point: 49-51°; Boiling Point: 231-232°; Flash point: 91°C (195°F)
Density (g/mL): 0.917