Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

α-Bromo-3-fluorotoulene ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

130130-25G 130130-50G 130130-5G
BT130130-25GEA 110 USD InStock
BT130130-25G BT130130-50G BT130130-5G
α-Bromo-3-fluorotoulene ≥98%
α-Bromo-3-fluorotoulene
CAS: 456-41-7; EC No: 207-263-2; MDL No: MFCD00000340 UN No: UN3265; Haz Class: 8; Packing Group: III Liquid; Molecular Formula: C7H6BrF ; MW: 189.02 Boiling Point: 88°/20 mmHg; Flash point: 62°C (144°F) Density (g/mL): 1.541; Refractive Index: 1.546

  • Refractive index: 1.546
  • Liquid


Formula: C₇H₆BrF
Boiling Pt: 88 °C/20 mmHg
Density: 1.541 g/mL
Flash Pt: 62 °C (144 °F)
MDL Number: MFCD00000340
CAS Number: 456-41-7
EINECS: 207-263-2
UN: 3265
ADR: 8,III
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty