Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hexafluorosilicic acid 23% (w/w) in water

Supplier: BEANTOWN CHEMICAL - BTC

130260-100G 130260-2.5KG 130260-500G
BT130260-100GEA 27 USD InStock
BT130260-100G BT130260-2.5KG BT130260-500G
Hexafluorosilicic acid 23% (w/w) in water
Hexafluorosilicic acid
CAS: 16961-83-4; EC No: 241-034-8; MDL No: MFCD00036289; RTECS: VV8225000 UN No: UN1778; Haz Class: 8; Packing Group: II Liquid; Linear Formula: H2SiF6; MW: 144.09 Density (g/mL): 1.22

  • RTECS: VV8225000
  • Liquid


Formula: H₂F₆Si
MW: 144.09 g/mol
Density: 1.22 g/mL
MDL Number: MFCD00036289
CAS Number: 16961-83-4
EINECS: 241-034-8
UN: 1778
ADR: 8 II
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty