Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diphenhydramine hydrochloride ≥99%

Supplier: BEANTOWN CHEMICAL - BTC

131320-1KG 131320-250G 131320-50G
BT131320-1KGEA 318 USD InStock
BT131320-1KG BT131320-250G BT131320-50G
Diphenhydramine hydrochloride ≥99%
Diphenhydramine hydrochloride
CAS: 147-24-0; EC No: 205-687-2; MDL No: MFCD00012479 UN No: UN2811; Haz Class: 6.1; Packing Group: III Solid; Linear Formula: (C6H5)2CHOCH2CH2N(CH3)2·HCl; Molecular Formula: C17H21NO·HCl; MW: 291.82 Light Sensitive

  • Light Sensitive
  • Solid


Formula: C₁₇H₂₁NO
MDL Number: MFCD00012479
CAS Number: 147-24-0
EINECS: 205-687-2
ADR: 6.1,III
Order Now
Specification Test Results
Assay 99%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty