Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromobutyronitrile ≥97%

Supplier: BEANTOWN CHEMICAL - BTC

131350-10G 131350-250G 131350-50G
BT131350-10GEA 53 USD InStock
BT131350-10G BT131350-250G BT131350-50G
4-Bromobutyronitrile ≥97%
4-Bromobutyronitrile
CAS: 5332-06-9; EC No: 226-236-6; MDL No: MFCD00001971; RTECS: ET8925000 UN No: UN3276; Haz Class: 6.1; Packing Group: III Liquid; Linear Formula: Br(CH2)3CN; MW: 148.01 Boiling Point: 205°; Flash point: 103°C (217°F) Density (g/mL): 1.489; Refractive Index: 1.478

  • RTECS: ET8925000
  • Refractive index: 1.478
  • Liquid


Formula: BrH₂CCH₂CH₂CN
Boiling Pt: 205 °C
Density: 1.489 g/mL
Flash Pt: 103 °C (217 °F)
MDL Number: MFCD00001971
CAS Number: 5332-06-9
EINECS: 226-236-6
UN: 3276
ADR: 6.1,III
Order Now
Specification Test Results
Assay 97%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty