Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl acetate ≥99.5% for HPLC

Supplier: BEANTOWN CHEMICAL - BTC

132480-1L 132480-4L 132480-4X1L
BT132480-1LEA 61.48 USD InStock
BT132480-1L BT132480-4L BT132480-4X1L
Ethyl acetate ≥99.5% for HPLC
Ethyl acetate
CAS: 141-78-6; EC No: 205-500-4; MDL No: MFCD00009171; RTECS: AH5425000 UN No: UN1173; Haz Class: 3; Packing Group: II Liquid; Linear Formula: CH3COOCH2CH3; Molecular Formula: C4H8O2; MW: 88.11 Melting Point: -84°; Boiling Point: 76.5-77.5°; Flash point: -3°C (27°F) Density (g/mL): 0.902; Refractive Index: 1.372

  • RTECS: AH5425000
  • Refractive index: 1.372
  • Liquid


Formula: CH₃COOC₂H₅
Boiling Pt: 76.5…77.5 °C
Melting Pt: –84 °C
Density: 0.902 g/mL
Flash Pt: –3 °C (27 °F)
MDL Number: MFCD00009171
CAS Number: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=0f6c525db7a2f3725affe5045040d70e0ac99352&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Assay 99.5% min.

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty