Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium hydride 57 - 63% dispersion in mineral oil

Supplier: BEANTOWN CHEMICAL - BTC

132755-250G 132755-50G
BT132755-250GEA 219.1 USD InStock
BT132755-250G BT132755-50G
Sodium hydride 57 - 63% dispersion in mineral oil
Sodium hydride
CAS: 7646-69-7; EC No: 231-587-3; MDL No: MFCD00003471 UN No: UN1427; Haz Class: 4.3; Packing Group: I Dispersion; Linear Formula: NaH; MW: 24.00 Melting Point: 800° (decomposes) Density (g/mL): 0.92 Air Sensitive, Moisture Sensitive

  • Air Sensitive, Moisture Sensitive
  • Dispersion


Formula: HNa
Melting Pt: 800 °C (dec.)
Density: 0.92 g/mL
MDL Number: MFCD00003471
CAS Number: 7646-69-7
EINECS: 231-587-3
UN: 1427
ADR: 4.3,I
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty