Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium tetrachloroaluminate

Supplier: BEANTOWN CHEMICAL - BTC

132955-100G 132955-25G 132955-5G
BT132955-100GEA 755 USD InStock
BT132955-100G BT132955-25G BT132955-5G
Sodium tetrachloroaluminate
Sodium tetrachloroaluminate
CAS: 7784-16-9; EC No: 232-050-6; MDL No: MFCD00050361 UN No: UN3260; Haz Class: 8; Packing Group: III Powder; Linear Formula: NaAlCl4; MW: 191.78 Melting Point: 185° Density (g/mL): 2.01 Moisture Sensitive

  • Moisture Sensitive
  • Powder


Formula: AlCl₄Na
MW: 191.78
Melting Pt: 154
Density: 2.01 g/mL
MDL Number: MFCD00050361
CAS Number: 7784-16-9
EINECS: 232-050-6
UN: 3260
ADR: 8 III
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty