Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ammonium hexachlororuthenate(IV)

Supplier: BEANTOWN CHEMICAL - BTC

133160-1G 133160-5G
BT133160-1GEA 65 USD InStock
BT133160-1G BT133160-5G
Ammonium hexachlororuthenate(IV)
Ammonium hexachlororuthenate(IV)
CAS: 18746-63-9; EC No: 242-552-7; MDL No: MFCD00015962 UN No: UN3288; Haz Class: 6.1; Packing Group: III Crystalline/Powder; Linear Formula: (NH4)2RuCl6; MW: 349.86 Hygroscopic

  • Hygroscopic
  • Crystalline/Powder


Formula: H₈Cl₆N₂Ru
MW: 349.86
MDL Number: MFCD00015962
CAS Number: 18746-63-9
EINECS: 242-552-7
ADR: 6.1 III
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty