Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium hydroxide 40% (w/v) in water

Supplier: BEANTOWN CHEMICAL - BTC

133415-20L 133415-4L
BT133415-20LEA 261.85 USD InStock
BT133415-20L BT133415-4L
Sodium hydroxide 40% (w/v) in water
Sodium hydroxide
CAS: 1310-73-2; EC No: 215-185-5; MDL No: MFCD00003548 UN No: UN1824; Haz Class: 8; Packing Group: II Liquid; Linear Formula: NaOH; MW: 40.00 Melting Point: -12-10°; Boiling Point: 105-140°

  • Liquid


Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390
Melting Pt: 323
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8 II
Merck Index: 14,08627
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty