Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,4-Butanediol dimethanesulfonate ≥98%

Supplier: BEANTOWN CHEMICAL - BTC
Danger

133510-25G 133510-5G
BT133510-25GEA 74 USD InStock
BT133510-25G BT133510-5G
1,4-Butanediol dimethanesulfonate ≥98%
Busulfan
CAS: 55-98-1; EC No: 200-250-2; MDL No: MFCD00007562; RTECS: EK1750000 UN No: UN2811; Haz Class: 6.1; Packing Group: II Crystalline/Solid; Molecular Formula: C6H14O6S2; MW: 246.30 Melting Point: 114-117°

  • RTECS: EK1750000


Formula: C₆H₁₄O₆S₂
MW: 246.29 g/mol
Melting Pt: 114…117 °C
Storage Temperature: Ambient
MDL Number: MFCD00007562
CAS Number: 55-98-1
EINECS: 200-250-2
UN: 2811
ADR: 6.1 II
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty