Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ammonium metavanadate ≥99.995%

Supplier: BEANTOWN CHEMICAL - BTC

133745-25G 133745-5G
BT133745-25GEA 274 USD InStock
BT133745-25G BT133745-5G
Ammonium metavanadate ≥99.995%
Ammonium metavanadate
CAS: 7803-55-6; EC No: 232-261-3; MDL No: MFCD00011430 UN No: UN2859; Haz Class: 6.1; Packing Group: II Powder; Linear Formula: NH4VO3; MW: 116.98 Density (g/mL): 2.32

  • Powder


Formula: NH₄VO₃
MW: 116.98 g/mol
Melting Pt: 200
Density: 2.32 g/mL
MDL Number: MFCD00011430
CAS Number: 7803-55-6
EINECS: 232-261-3
UN: 2859
ADR: 6.1 II
Merck Index: 13,00572
Order Now
Specification Test Results
Assay 99.995%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty