134120-100G
134120-25G
134120-500G
BT134120-100GEA
136
USD
InStock
BT134120-100G
BT134120-25G
BT134120-500G
Sodium hexafluoroaluminate ≥99.9% (trace metals basis)
Sodium hexafluoroaluminate
CAS: 13775-53-6; EC No: 237-410-6; MDL No: MFCD00003507; RTECS: BD0075000
UN No: UN3260; Haz Class: 8; Packing Group: III
Powder; Linear Formula: Na3AlF6; MW: 209.95
Melting Point: 1000°
Density (g/mL): 2.90