134270-1G
134270-25G
134270-5G
BT134270-1GEA
26
USD
InStock
BT134270-1G
BT134270-25G
BT134270-5G
Antimony ≥99.999% (trace metals basis), powder -200 mesh
Antimony
CAS: 7440-36-0; EC No: 231-146-5; MDL No: MFCD00134030; RTECS: CC4025000
UN No: UN2871; Haz Class: 6.1; Packing Group: III
Powder; Molecular Formula: Sb; MW: 121.76
Melting Point: 630°; Boiling Point: 1635°
Density (g/mL): 6.69