Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hafnium(IV) fluoride ≥99.9%

Supplier: BEANTOWN CHEMICAL - BTC

134415-25G 134415-5G
BT134415-25GEA 612 USD InStock
BT134415-25G BT134415-5G
Hafnium(IV) fluoride ≥99.9%
Hafnium(IV) fluoride
CAS: 13709-52-9; EC No: 237-258-0; MDL No: MFCD00016127 UN No: UN3290; Haz Class: 6.1 (8); Packing Group: II -60 Mesh Powder; Linear Formula: HfF4; MW: 254.48 Moisture Sensitive

  • Moisture Sensitive
  • -60 Mesh Powder


Formula: F₄Hf
MW: 254.48
MDL Number: MFCD00016127
CAS Number: 13709-52-9
EINECS: 237-258-0
UN: 3290
ADR: 6.1 II
Order Now
Specification Test Results
Assay 99.9%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty