134595-100G
134595-25G
134595-5G
BT134595-100GEA
452
USD
InStock
BT134595-100G
BT134595-25G
BT134595-5G
Triisopropylsilyl trifluoromethanesulphonate ≥97%
Triisopropylsilyl trifluoromethanesulphonate
CAS: 80522-42-5; MDL No: MFCD00009913
UN No: UN3265; Haz Class: 8; Packing Group: II
Liquid; Linear Formula: CF3SO3Si[CH(CH3)2]3; Molecular Formula: C10H21F3O3SSi; MW: 306.42
Boiling Point: 45-46°/0.03 mmHg; Flash point: 100°C (212°F)
Density (g/mL): 1.136; Refractive Index: 1.415
Moisture Sensitive
- Refractive index: 1.415
- Moisture Sensitive
- Liquid