Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Imidazole ≥99%

Supplier: BEANTOWN CHEMICAL - BTC

134805-100G 134805-2.5KG 134805-500G
BT134805-100GEA 36.73 USD InStock
BT134805-100G BT134805-2.5KG BT134805-500G
Imidazole ≥99%
Imidazole
CAS: 288-32-4; EC No: 206-019-2; MDL No: MFCD00005183; RTECS: NI3325000 UN No: UN2923; Haz Class: 8 (6.1); Packing Group: III Crystalline; Molecular Formula: C3H4N2; MW: 68.08 Melting Point: 85-92°; Boiling Point: 256°; Flash point: 145°C (293°F)

  • RTECS: NI3325000
  • Crystalline


Formula: C₃H₄N₂
MW: 68.08 g/mol
Boiling Pt: 255
Melting Pt: 85…92 °C
Flash Pt: 145 °C (293 °F)
MDL Number: MFCD00005183
CAS Number: 288-32-4
EINECS: 206-019-2
UN: 2923
ADR: 8 III
Merck Index: 13,04935
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48eba49e61201d6fffa2bef0c54633a20660c7ef&vid=ddb38a8e40ebda2af69627b5ed326af620745f06&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Assay 99%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty