Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Butyl formate ≥97%

Supplier: BEANTOWN CHEMICAL - BTC

134905-1L 134905-250ML 134905-50ML
BT134905-1LEA 223 USD InStock
BT134905-1L BT134905-250ML BT134905-50ML
Butyl formate ≥97%
Butyl formate
CAS: 592-84-7; EC No: 209-772-5; MDL No: MFCD00003296; RTECS: LQ5500000 UN No: UN1128; Haz Class: 3; Packing Group: II Liquid; Linear Formula: CH3(CH2)3OCHO; Molecular Formula: C5H10O2; MW: 102.13 Melting Point: -92°; Boiling Point: 106-107°; Flash point: 14°C (57°F) Density (g/mL): 0.892; Refractive Index: 1.389

  • RTECS: LQ5500000
  • Refractive index: 1.389
  • Liquid


Formula: C₅H₁₀O₂
Boiling Pt: 106…107 °C
Melting Pt: –92 °C
Density: 0.892 g/mL
Flash Pt: 14 °C (57 °F)
MDL Number: MFCD00003296
CAS Number: 592-84-7
EINECS: 209-772-5
UN: 1128
ADR: 3,II
Order Now
Specification Test Results
Assay 97%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty