Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,2-Dibromo-1-phenylethane ≥97%

Supplier: BEANTOWN CHEMICAL - BTC

135010-100G 135010-25G 135010-500G
BT135010-100GEA 77 USD InStock
BT135010-100G BT135010-25G BT135010-500G
1,2-Dibromo-1-phenylethane ≥97%
1,2-Dibromo-1-phenylethane
CAS: 93-52-7; EC No: 202-253-4; MDL No: MFCD00000138 UN No: UN3261; Haz Class: 8; Packing Group: III Crystalline; Linear Formula: C6H5CH(Br)CH2Br; Molecular Formula: C8H8Br2 ; MW: 263.96 Melting Point: 70-74°; Boiling Point: 139-141°/15 mmHg

  • Crystalline


Formula: C₆H₅CHBrCH₂Br
MW: 263.96 g/mol
Boiling Pt: 139…141 °C/15 mmHg
Melting Pt: 70…74 °C
MDL Number: MFCD00000138
CAS Number: 93-52-7
EINECS: 202-253-4
UN: 3261
ADR: 8 III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee6d6874d48c69b9de8b135a68f7f21d6b917e29&vid=ee6d6874d48c69b9de8b135a68f7f21d6b917e29&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Assay 97%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty