Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Selenium tetrachloride ≥99.5% (trace metals basis)

Supplier: BEANTOWN CHEMICAL - BTC

135190-100G 135190-25G
BT135190-100GEA 476 USD InStock
BT135190-100G BT135190-25G
Selenium tetrachloride ≥99.5% (trace metals basis)
Selenium tetrachloride
CAS: 10026-03-6; EC No: 233-053-5; MDL No: MFCD00011225 UN No: UN2923; Haz Class: 8 (6.1); Packing Group: II Lump; Linear Formula: SeCl4; MW: 220.77 Melting Point: 170-196°; Boiling Point: 288° Density (g/mL): 2.6; Refractive Index: 1.807 Moisture Sensitive

  • Refractive index: 1.807
  • Moisture Sensitive
  • Lump


Formula: Cl₄Se
Boiling Pt: 288 °C
Melting Pt: 170…196 °C
Density: 2.6 g/mL
MDL Number: MFCD00011225
CAS Number: 10026-03-6
EINECS: 233-053-5
UN: 2923
ADR: 8,II
Order Now
Specification Test Results
Assay 99.5%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty