Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetrahydrothiophene ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

135360-100G 135360-500G
BT135360-100GEA 30.59 USD InStock
BT135360-100G BT135360-500G
Tetrahydrothiophene ≥98%
Tetrahydrothiophene
CAS: 110-01-0; EC No: 203-728-9; MDL No: MFCD00005476; RTECS: XN0370000 UN No: UN2412; Haz Class: 3; Packing Group: II Liquid; Molecular Formula: C4H8S; MW: 88.17 Melting Point: -96°; Boiling Point: 120-121°; Flash point: 13°C (55°F) Density (g/mL): 0.999; Refractive Index: 1.504

  • RTECS: XN0370000
  • Refractive index: 1.504
  • Liquid


Formula: C₄H₈S
Boiling Pt: 120…121 °C
Melting Pt: –96 °C
Density: 0.999 g/mL
Flash Pt: 13 °C (55 °F)
MDL Number: MFCD00005476
CAS Number: 110-01-0
EINECS: 203-728-9
UN: 2412
ADR: 3,II
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty