Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl bromoacetate ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

135900-100G 135900-500G
BT135900-100GEA 52.29 USD InStock
BT135900-100G BT135900-500G
Ethyl bromoacetate ≥98%
Ethyl bromoacetate
CAS: 105-36-2; EC No: 203-290-9; MDL No: MFCD00000191; RTECS: AF6000000 UN No: UN1603; Haz Class: 6.1 (3); Packing Group: II Liquid; Linear Formula: BrCH2COOCH2CH3; Molecular Formula: C4H7BrO2; MW: 167.00 Boiling Point: 159°; Flash point: 47°C (117°F) Density (g/mL): 1.506; Refractive Index: 1.451

  • RTECS: AF6000000
  • Refractive index: 1.451
  • Liquid


Formula: BrCH₂COOCH₂CH₃
MW: 167 g/mol
Boiling Pt: 159
Melting Pt: -38
Density: 1.506 g/mL
Flash Pt: 47 °C (117 °F)
MDL Number: MFCD00000191
CAS Number: 105-36-2
EINECS: 203-290-9
UN: 1603
ADR: 6.1 II
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty