Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ammonium hexafluorophosphate ≥99.5%

Supplier: BEANTOWN CHEMICAL - BTC

136385-100G 136385-25G
BT136385-100GEA 174 USD InStock
BT136385-100G BT136385-25G
Ammonium hexafluorophosphate ≥99.5%
Ammonium hexafluorophosphate
CAS: 16941-11-0; EC No: 241-009-1; MDL No: MFCD00064642 UN No: UN3260; Haz Class: 8; Packing Group: III Powder; Linear Formula: NH4PF6; MW: 163.00 Density (g/mL): 2.18 Hygroscopic

  • Hygroscopic
  • Powder


Formula: H₄F₆NP
MW: 163 g/mol
Melting Pt: 198
Density: 2.18 g/mL
MDL Number: MFCD00064642
CAS Number: 16941-11-0
EINECS: 241-009-1
UN: 3260
ADR: 8 III
Merck Index: 12,00557
Order Now
Specification Test Results
Assay 99.5%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty