Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-1,1,1-trifluoroacetone ≥97%

Supplier: BEANTOWN CHEMICAL - BTC

136425-100G 136425-25G 136425-5G
BT136425-100GEA 613 USD InStock
BT136425-100G BT136425-25G BT136425-5G
3-Bromo-1,1,1-trifluoroacetone ≥97%
3-Bromo-1,1,1-trifluoroacetone
CAS: 431-35-6; EC No: 207-071-9; MDL No: MFCD00039237 UN No: UN2924; Haz Class: 3(8); Packing Group: II Liquid; Linear Formula: BrCH2COCF3; MW: 190.95 Boiling Point: 87°/743 mmHg; Flash point: 5°C (41°F) Density (g/mL): 1.839; Refractive Index: 1.376

  • Refractive index: 1.376
  • Liquid


Formula: BrH₂CC(O)CF₃
Boiling Pt: 87 °C/743 mmHg
Density: 1.839 g/mL
Flash Pt: 5 °C (41 °F)
MDL Number: MFCD00039237
CAS Number: 431-35-6
EINECS: 207-071-9
UN: 2924
ADR: 3,II
Order Now
Specification Test Results
Assay 97%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty