Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Samarium ≥99.9% (trace rare earth metals basis), pieces, sublimed dendritic

Supplier: BEANTOWN CHEMICAL - BTC

137480-10G 137480-250G 137480-50G
BT137480-10GEA 56.06 USD InStock
BT137480-10G BT137480-250G BT137480-50G
Samarium ≥99.9% (trace rare earth metals basis), pieces, sublimed dendritic
Samarium
CAS: 7440-19-9; EC No: 231-128-7; MDL No: MFCD00011233 UN No: UN3178; Haz Class: 4.1; Packing Group: II Solid; Molecular Formula: Sm; MW: 150.36 Melting Point: 1074°; Boiling Point: 1794° Density (g/mL): 7.47

  • Solid


Formula: Sm
Boiling Pt: 1794 °C
Melting Pt: 1074 °C
Density: 7.47 g/mL
MDL Number: MFCD00011233
CAS Number: 7440-19-9
EINECS: 231-128-7
ADR: 4.1,II
Order Now
Specification Test Results
Assay 99.9%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty