137775-1G
137775-25G
137775-5G
BT137775-1GEA
21
USD
InStock
BT137775-1G
BT137775-25G
BT137775-5G
2-Isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98%
2-Isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
CAS: 61676-62-8; MDL No: MFCD00192241
UN No: UN1993; Haz Class: 3; Packing Group: III
Liquid; Molecular Formula: C9H19BO3; MW: 186.06
Boiling Point: 73°/15 mmHg; Flash point: 43°C (109°F)
Density (g/mL): 0.912; Refractive Index: 1.409
Moisture Sensitive
- Refractive index: 1.409
- Moisture Sensitive
- Liquid