Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Copper(II) nitrate hemipentahydrate 98.0-102.0% ACS

Supplier: BEANTOWN CHEMICAL - BTC

137915-100G 137915-2KG 137915-500G
BT137915-100GEA 39.16 USD InStock
BT137915-100G BT137915-2KG BT137915-500G
Copper(II) nitrate hemipentahydrate 98.0-102.0% ACS
Copper(II) nitrate hemipentahydrate
CAS: 19004-19-4; EC No: 221-838-5; MDL No: MFCD00149671; RTECS: GL7875000 UN No: UN3085; Haz Class: 5.1 (8); Packing Group: III Crystalline/Lump; Linear Formula: Cu(NO3)2·2.5H2O; MW: 232.59 Density (g/mL): 2.32 Hygroscopic

  • RTECS: GL7875000
  • Hygroscopic
  • Crystalline/Lump


Formula: Cu(NO₃)₂·2.5H₂O
MW: 232.59 g/mol
Melting Pt: 255
Density: 2.32 g/mL
MDL Number: MFCD02253001
CAS Number: 19004-19-4
EINECS: 221-838-5
UN: 3085
ADR: 5.1 III
Order Now
Specification Test Results
Assay 98.0-102.0%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty