Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Silicon ≥99.995% (trace metals basis), powder, crystalline amorphous 1-20 µm

Supplier: BEANTOWN CHEMICAL - BTC

138010-250G 138010-50G
BT138010-250GEA 324 USD InStock
BT138010-250G BT138010-50G
Silicon ≥99.995% (trace metals basis), powder, crystalline amorphous 1-20 µm
Silicon
CAS: 7440-21-3; EC No: 231-130-8; MDL No: MFCD00085311; RTECS: VW0400000 UN No: UN1346; Haz Class: 4.1; Packing Group: III Powder; Molecular Formula: Si; MW: 28.09 Melting Point: 1410°; Boiling Point: 2355° Density (g/mL): 2.33

  • RTECS: VW0400000
  • Powder


Formula: Si
MW: 28.09 g/mol
Boiling Pt: 2355 °C
Melting Pt: 1410 °C
Density: 2.33 g/mL
MDL Number: MFCD00085311
CAS Number: 7440-21-3
EINECS: 231-130-8
UN: 1346
ADR: 4.1 III
Merck Index: 12,08635
Order Now
Specification Test Results
Assay 99.995%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty