138180-10G
138180-250G
138180-50G
BT138180-10GEA
18
USD
InStock
BT138180-10G
BT138180-250G
BT138180-50G
Benzyldimethylhexadecylammonium chloride hydrate ≥95%
Benzyldimethylhexadecylammonium chloride hydrate
CAS: 122-18-9; EC No: 204-526-3; MDL No: MFCD00011625; RTECS: BO6822450
UN No: UN3261; Haz Class: 8; Packing Group: II
Powder; Molecular Formula: C25H46ClN·xH2O; MW: 396.09 (anhydrous)