Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Barium fluoride ≥99.99%

Supplier: BEANTOWN CHEMICAL - BTC

138205-100G 138205-25G 138205-5G
BT138205-100GEA 493 USD InStock
BT138205-100G BT138205-25G BT138205-5G
Barium fluoride ≥99.99%
Barium fluoride
CAS: 7787-32-8; EC No: 232-108-0; MDL No: MFCD00003450 UN No: UN1564; Haz Class: 6.1; Packing Group: III Powder; Molecular Formula: BaF2; MW: 175.32 Boiling Point: 2260° Density (g/mL): 4.89

  • Powder


Formula: BaF₂
MW: 175.33 g/mol
Boiling Pt: 2260
Melting Pt: 1270
Density: 4.89 g/mL
MDL Number: MFCD00003450
CAS Number: 7787-32-8
EINECS: 232-108-0
UN: 1564
ADR: 6.1 III
Merck Index: 12,01003
Order Now
Specification Test Results
Assay 99.99%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty