Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Palladium, black ≥99.9%

Supplier: BEANTOWN CHEMICAL - BTC

138220-1G 138220-5G
BT138220-1GEA 188 USD InStock
BT138220-1G BT138220-5G
Palladium, black ≥99.9%
Palladium
CAS: 7440-05-3; EC No: 231-115-6; MDL No: MFCD00011167; RTECS: RT3480500 UN No: UN3089; Haz Class: 4.1; Packing Group: II Solid, S.A. typically 20 m2/g; Molecular Formula: Pd; MW: 106.42 Melting Point: 1554°; Boiling Point: 2970° Density (g/mL): 12.02

  • RTECS: RT3480500
  • Solid, S.A. typically 20 m²/g


Formula: Pd
MW: 106.420 g/mol
Boiling Pt: 2970 °C
Melting Pt: 1554 °C
Density: 12.02 g/mL
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6
ADR: 4.1 II
Order Now
Specification Test Results
Assay 99.9%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty