Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

trans-4-Octene ≥97%

Supplier: BEANTOWN CHEMICAL - BTC

138915-25G 138915-5G
BT138915-25GEA 213.95 USD InStock
BT138915-25G BT138915-5G
trans-4-Octene ≥97%
trans-4-Octene
CAS: 14850-23-8; EC No: 238-913-3; MDL No: MFCD00009476 UN No: UN3295; Haz Class: 3; Packing Group: II Liquid; Linear Formula: CH3CH2CH2CH=CHCH2CH2CH3; Molecular Formula: C8H16; MW: 112.22 Melting Point: -94°; Boiling Point: 122-123°; Flash point: 21°C (70°F) Density (g/mL): 0.715; Refractive Index: 1.412

  • Refractive index: 1.412
  • Liquid


Formula: C₈H₁₆
MW: 112.22 g/mol
Boiling Pt: 122
Melting Pt: -93.8
Density: 0.715 g/mL
Flash Pt: 21 °C (70 °F)
MDL Number: MFCD00009476
CAS Number: 14850-23-8
EINECS: 238-913-3
UN: 3295
ADR: 3 II
Order Now
Specification Test Results
Assay 97%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty