Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Glutaraldehyde 50% (w/w) in water

Supplier: BEANTOWN CHEMICAL - BTC

139070-100G 139070-2.5KG 139070-500G
BT139070-100GEA 27.45 USD InStock
BT139070-100G BT139070-2.5KG BT139070-500G
Glutaraldehyde 50% (w/w) in water
Glutaraldehyde
CAS: 111-30-8; EC No: 203-856-5; MDL No: MFCD00007025; RTECS: MA2450000 UN No: UN2922; Haz Class: 8 (6.1); Packing Group: III Liquid; Linear Formula: OHC(CH2)3CHO; Molecular Formula: C5H8O2; MW: 100.12 Density (g/mL): 1.106

  • RTECS: MA2450000
  • Liquid


Formula: C₅H₈O₂
Density: 1.106 g/mL
MDL Number: MFCD00007025
CAS Number: 111-30-8
EINECS: 203-856-5
UN: 2922
ADR: 6.1,III
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty