Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Aluminium isopropoxide ≥99.99%

Supplier: BEANTOWN CHEMICAL - BTC

139895-100G 139895-25G
BT139895-100GEA 285 USD InStock
BT139895-100G BT139895-25G
Aluminium isopropoxide ≥99.99%
Aluminium isopropoxide
CAS: 555-31-7; EC No: 209-090-8; MDL No: MFCD00008870; RTECS: BD0975000 UN No: UN3181; Haz Class: 4.1; Packing Group: II Powder; Linear Formula: Al[OCH(CH3)2]3 ; MW: 204.24 Melting Point: 128-133° Density (g/mL): 1.035 Moisture Sensitive

  • RTECS: BD0975000
  • Moisture Sensitive
  • Powder


Formula: C₉H₂₁AlO₃
MW: 204.25 g/mol
Boiling Pt: 135
Melting Pt: 119
Density: 1.035 g/mL
MDL Number: MFCD00008870
CAS Number: 555-31-7
EINECS: 209-090-8
UN: 3181
ADR: 4.1 II
Merck Index: 13,00346
Order Now
Specification Test Results
Assay 99.99%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty