Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Triphenyl phosphite ≥97%

Supplier: BEANTOWN CHEMICAL - BTC

140035-2.5KG 140035-500G
BT140035-2.5KGEA 124 USD InStock
BT140035-2.5KG BT140035-500G
Triphenyl phosphite ≥97%
Triphenyl phosphite
CAS: 101-02-0; EC No: 202-908-4; MDL No: MFCD00003032 UN No: UN3077; Haz Class: 9; Packing Group: III Liquid; Linear Formula: (C6H5O)3P; Molecular Formula: C18H15O3P; MW: 310.29 Melting Point: 22-24°; Boiling Point: 360°; Flash point: 146°C (295°F) Density (g/mL): 1.184; Refractive Index: 1.590 Air Sensitive, Moisture Sensitive

  • Refractive index: 1.590
  • Air Sensitive, Moisture Sensitive
  • Liquid


Formula: C₁₈H₁₅O₃P
MW: 310.29 g/mol
Boiling Pt: 360
Melting Pt: 22…24 °C
Density: 1.184 g/mL
Flash Pt: 146 °C (295 °F)
MDL Number: MFCD00003032
CAS Number: 101-02-0
EINECS: 202-908-4
ADR: 9 III
Order Now
Specification Test Results
Assay 97%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty