140160-1G
140160-5G
BT140160-1GEA
145.05
USD
InStock
BT140160-1G
BT140160-5G
Scandium(III) nitrate hydate ≥99.99%
Scandium(III) nitrate hydrate
CAS: 13465-60-6; EC No: 236-701-5; MDL No: MFCD00011220
UN No: UN1477; Haz Class: 5.1; Packing Group: III
Crystalline Aggregates; Linear Formula: Sc(NO3)3·xH2O; MW: 230.97 (anhydrous)
Hygroscopic
- Crystalline Aggregates
- Hygroscopic