Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hexamethylcyclotrisiloxane ≥97%

Supplier: BEANTOWN CHEMICAL - BTC

140240-100G 140240-25G 140240-500G
BT140240-100GEA 101.06 USD InStock
BT140240-100G BT140240-25G BT140240-500G
Hexamethylcyclotrisiloxane ≥97%
Hexamethylcyclotrisiloxane
CAS: 541-05-9; EC No: 208-765-4; MDL No: MFCD00005943; RTECS: GZ4705000 UN No: UN1325; Haz Class: 4.1; Packing Group: II Crystalline; Molecular Formula: C6H18O3Si3; MW: 222.47 Melting Point: 50-64°; Boiling Point: 134°; Flash point: 35°C (95°F) Moisture Sensitive

  • RTECS: GZ4705000
  • Moisture Sensitive
  • Crystalline


Formula: C₆H₁₈O₃Si₃
MW: 222.46 g/mol
Boiling Pt: 134
Melting Pt: 64.5
Flash Pt: 35 °C (95 °F)
MDL Number: MFCD00005943
CAS Number: 541-05-9
EINECS: 208-765-4
UN: 1325
ADR: 4.1 II
Order Now
Specification Test Results
Assay 97%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty