Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Octamethylcyclotetrasiloxane ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

140305-100G 140305-500G
BT140305-100GEA 73.93 USD InStock
BT140305-100G BT140305-500G
Octamethylcyclotetrasiloxane ≥98%
Octamethylcyclotetrasiloxane
CAS: 556-67-2; EC No: 209-136-7; MDL No: MFCD00003269; RTECS: GZ4397000 UN No: UN1993; Haz Class: 3; Packing Group: III Liquid; Linear Formula: [-Si(CH3)2O-]4; Molecular Formula: C8H24O4Si4; MW: 296.62 Melting Point: 71-18°; Boiling Point: 175-176°; Flash point: 55°C (131°F) Density (g/mL): 0.956; Refractive Index: 1.396 Moisture Sensitive

  • RTECS: GZ4397000
  • Refractive index: 1.396
  • Moisture Sensitive
  • Liquid


Formula: [-Si(CH₃)₂O-]₄
Boiling Pt: 175…176 °C
Melting Pt: 71…18 °C
Density: 0.956 g/mL
Flash Pt: 55 °C (131 °F)
MDL Number: MFCD00003269
CAS Number: 556-67-2
EINECS: 209-136-7
UN: 1993
ADR: 3,III
Order Now
Specification Test Results
Assay 98%

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty