Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromoisobutyric acid ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

140325-100G 140325-25G 140325-500G
BT140325-100GEA 79 USD InStock
BT140325-100G BT140325-25G BT140325-500G
2-Bromoisobutyric acid ≥98%
2-Bromoisobutyric acid
CAS: 2052-01-9; EC No: 218-139-2; MDL No: MFCD00004168 UN No: UN3261; Haz Class: 8; Packing Group: III Powder; Molecular Formula: C4H7BrO2 ; MW: 167.00 Melting Point: 44-47°; Boiling Point: 198-200°; Flash point: 110°C (230°F)

  • Powder


Formula: C₄H₇BrO₂
MW: 167 g/mol
Boiling Pt: 198…200 °C
Melting Pt: 48
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00004168
CAS Number: 2052-01-9
EINECS: 218-139-2
UN: 3261
ADR: 8 III
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty