Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Biphenylcarbonyl chloride ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

140690-10G 140690-250G 140690-50G
BT140690-10GEA 56.54 USD InStock
BT140690-10G BT140690-250G BT140690-50G
4-Biphenylcarbonyl chloride ≥98%
4-Biphenylcarbonyl chloride
CAS: 14002-51-8; EC No: 237-804-8; MDL No: MFCD00000692 UN No: UN3261; Haz Class: 8; Packing Group: II Powder; Molecular Formula: C13H9ClO; MW: 216.66 Melting Point: 110-112° Moisture Sensitive

  • Moisture Sensitive
  • Powder


Formula: C₁₃H₉ClO
MW: 216.67 g/mol
Melting Pt: 110…112 °C
MDL Number: MFCD00000692
CAS Number: 14002-51-8
EINECS: 237-804-8
UN: 3261
ADR: 8 II
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty