Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-4-(trifluoromethyl)pyridine ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

140705-1G 140705-250MG
BT140705-1GEA 202 USD InStock
BT140705-1G BT140705-250MG
3-Fluoro-4-(trifluoromethyl)pyridine ≥98%
3-Fluoro-4-(trifluoromethyl)pyridine
CAS: 113770-87-9; EC No: 000-000-0; MDL No: MFCD03789250 UN No: UN1993; Haz Class: 3; Packing Group: III Liquid; Molecular Formula: C6H3F4N; MW: 165.09 Boiling Point: 37°/10 mmHg; Flash point: 20°C (68°F) Density (g/mL): 1.391; Refractive Index: 1.406

  • Refractive index: 1.406
  • Liquid


Formula: C₆H₃F₄N
MW: 165.09 g/mol
Boiling Pt: 37 °C/10 mmHg
Density: 1.391 g/mL
Flash Pt: 20 °C (68 °F)
MDL Number: MFCD03789250
CAS Number: 113770-87-9
ADR: 3 III
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty