Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methylphosphonic acid ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

140835-1G 140835-25G 140835-5G
BT140835-1GEA 21 USD InStock
BT140835-1G BT140835-25G BT140835-5G
Methylphosphonic acid ≥98%
Methylphosphonic acid
CAS: 993-13-5; EC No: 213-607-2; MDL No: MFCD00002137 UN No: UN3261; Haz Class: 8; Packing Group: II Solid; Linear Formula: CH3P(O)(OH)2; Molecular Formula: CH5O3P; MW: 96.03 Melting Point: 105-107°

  • Solid


Formula: CH₅O₃P
MW: 96.02 g/mol
Melting Pt: 107.5
MDL Number: MFCD00002137
CAS Number: 993-13-5
EINECS: 213-607-2
UN: 3261
ADR: 8 II
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty