Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Propanol ≥99.5% ACS

Supplier: BEANTOWN CHEMICAL - BTC

141195-1L 141195-4L 141195-500ML
BT141195-1LEA 55.08 USD InStock
BT141195-1L BT141195-4L BT141195-500ML
1-Propanol ≥99.5% ACS
1-Propanol
CAS: 71-23-8; EC No: 200-746-9; MDL No: MFCD00002941; RTECS: UH8225000 UN No: UN1274; Haz Class: 3; Packing Group: II Liquid; Linear Formula: CH3CH2CH2OH; Molecular Formula: C3H8; MW: 60.10 Melting Point: -127°; Boiling Point: 97°; Flash point: 22°C (72°F) Density (g/mL): 0.804; Refractive Index: 1.384

  • RTECS: UH8225000
  • Refractive index: 1.384
  • Liquid


Formula: H₃CCH₂CH₂OH
MW: 60.1 g/mol
Boiling Pt: 97
Melting Pt: -127
Density: 0.804 g/mL
Flash Pt: 22 °C (72 °F)
MDL Number: MFCD00002941
CAS Number: 71-23-8
EINECS: 200-746-9
UN: 1274
ADR: 3 II
Merck Index: 13,07934
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab459f809f7797b46eb6d275aa3b1e6e94dc70d1&vid=b4a45c3a7a92d9e6aed8d1d3bcf6b139b7a0e31f&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Assay 99.5% min.
Solubility in water Passes test
Color (APHA) ≤10
Residue after evaporation ≤0.001%
Water ≤0.2%
Titrable acid ≤0.0004meq/g
Ethyl alcohol ≤0.01%
Methanol ≤0.01%
Isopropyl alcohol ≤0.05%
Carbonyl compounds ≤0.03%
Liquid chromatography (ACS) Passes test
Gradient elution Passes test

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty