Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Selenic acid 40% (w/w) in water

Supplier: BEANTOWN CHEMICAL - BTC

141390-100G 141390-500G
BT141390-100GEA 183 USD InStock
BT141390-100G BT141390-500G
Selenic acid 40% (w/w) in water
Selenic acid
CAS: 7783-08-6; EC No: 231-979-4; MDL No: MFCD00012191 UN No: UN2922; Haz Class: 8 (6.1); Packing Group: II Liquid; Linear Formula: H2SeO4; MW: 144.97 Density (g/mL): 1.407

  • Liquid


Formula: H₂O₄Se
MW: 144.97
Boiling Pt: 260
Melting Pt: 58
Density: 2.6
MDL Number: MFCD00012191
CAS Number: 7783-08-6
EINECS: 231-979-4
UN: 2922
ADR: 8 II
Merck Index: 12,08570
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty