Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Trimethyl borate 99,9995

Supplier: BEANTOWN CHEMICAL - BTC

141565-10G 141565-250G 141565-50G
BT141565-10GEA 21 USD InStock
BT141565-10G BT141565-250G BT141565-50G
Trimethyl borate 99,9995
Trimethyl borate
CAS: 121-43-7; EC No: 204-468-9; MDL No: MFCD00008346; RTECS: ED5600000 UN No: UN2416; Haz Class: 3; Packing Group: II Liquid; Linear Formula: B(OCH3)3; Molecular Formula: C3H9BO3; MW: 103.92 Melting Point: -34°; Boiling Point: 68-69°; Flash point: -8°C (18°F) Density (g/mL): 0.932; Refractive Index: 1.346 Moisture Sensitive

  • RTECS: ED5600000
  • Refractive index: 1.346
  • Moisture Sensitive
  • Liquid


Formula: C₃H₉BO₃
MW: 103.91 g/mol
Boiling Pt: 67
Melting Pt: -5
Density: 0.932 g/mL
Flash Pt: –8 °C (18 °F)
MDL Number: MFCD00008346
CAS Number: 121-43-7
EINECS: 204-468-9
UN: 2416
ADR: 3 II
Merck Index: 12,09842
Order Now
Specification Test Results
Assay 99.9995%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty