141650-100G
141650-1KG
141650-25G
BT141650-100GEA
39.92
USD
InStock
BT141650-100G
BT141650-1KG
BT141650-25G
Sodium dithionite ≥85%, Technical Grade
Sodium dithionite
CAS: 7775-14-6; EC No: 231-890-0; MDL No: MFCD00011640
UN No: UN1384; Haz Class: 4.2; Packing Group: II
Powder; Linear Formula: NaO2SSO2Na; MW: 174.11
Melting Point: 300°
Density (g/mL): 2.5
Air Sensitive, Heat Sensitive, Moisture Sensitive
- Air Sensitive, Heat Sensitive, Moisture Sensitive
- Powder