Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium hydroxide ≥98%, soluble

Supplier: BEANTOWN CHEMICAL - BTC

142245-10KG 142245-2.5KG 142245-500G
BT142245-10KGEA 295.82 USD InStock
BT142245-10KG BT142245-2.5KG BT142245-500G
Sodium hydroxide ≥98%, soluble
Sodium hydroxide
CAS: 1310-73-2; EC No: 215-185-5; MDL No: MFCD00003548; RTECS: WB4900000 UN No: UN1823; Haz Class: 8; Packing Group: II Flakes; Linear Formula: NaOH; MW: 40.00 Melting Point: 318°; Boiling Point: 1390° Density (g/mL): 2.13 Air Sensitive , Hygroscopic

  • RTECS: WB4900000
  • Air Sensitive, Hygroscopic
  • Flakes


Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390
Melting Pt: 323
Density: 2.13 g/mL
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1823
ADR: 8 II
Merck Index: 14,08627
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty