Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-4-pentenoate ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

142385-1G 142385-25G 142385-5G
BT142385-1GEA 27.98 USD InStock
BT142385-1G BT142385-25G BT142385-5G
Ethyl-4-pentenoate ≥98%
Ethyl-4-pentenoate
CAS: 1968-40-7; EC No: 217-818-0; MDL No: MFCD00036562 UN No: UN3272; Haz Class: 3; Packing Group: III Liquid; Linear Formula: CH2=CHCH2CH2CO2CH2CH3; Molecular Formula: C7H12O2; MW: 128.17 Flash point: 37°C (98°F) Density (g/mL): 0.9

  • Liquid


Formula: C₇H₁₂O₂
MW: 128.17 g/mol
Density: 0.9 g/mL
Flash Pt: 37 °C (98 °F)
MDL Number: MFCD00036562
CAS Number: 1968-40-7
EINECS: 217-818-0
ADR: 3 III
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty