Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Lithium hexafluorosilicate

Supplier: BEANTOWN CHEMICAL - BTC

142640-25G 142640-5G
BT142640-25GEA 174 USD InStock
BT142640-25G BT142640-5G
Lithium hexafluorosilicate
Lithium hexafluorosilicate
CAS: 17347-95-4; EC No: 241-372-6; MDL No: MFCD00049473 UN No: UN2856; Haz Class: 6.1; Packing Group: III Powder; Linear Formula: Li2SiF6 ; MW: 155.96

  • Powder


Formula: F₆Li₂Si
MDL Number: MFCD00049473
CAS Number: 17347-95-4
EINECS: 241-372-6
UN: 2856
ADR: 6.1,III
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty