Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dibenzylamine ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

142705-1KG 142705-250G 142705-5KG
BT142705-1KGEA 120 USD InStock
BT142705-1KG BT142705-250G BT142705-5KG
Dibenzylamine ≥98%
Dibenzylamine
CAS: 103-49-1; EC No: 203-117-7; MDL No: MFCD00004770 UN No: UN2735; Haz Class: 8; Packing Group: III Liquid; Linear Formula: (C6H5CH2)2NH; Molecular Formula: C14H15N; MW: 197.28 Melting Point: -26°; Boiling Point: 300°; Flash point: 143°C (289°F) Density (g/mL): 1.026; Refractive Index: 1.574

  • Refractive index: 1.574
  • Liquid


Formula: C₆H₅CH₂NHCH₂C₆H₅
MW: 197.28 g/mol
Boiling Pt: 300
Melting Pt: -26
Density: 1.026 g/mL
Flash Pt: 143 °C (289 °F)
MDL Number: MFCD00004770
CAS Number: 103-49-1
EINECS: 203-117-7
UN: 2735
ADR: 6.1 III
Merck Index: 13,03035
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c17e97ec30680cc9e622a04bff55a48e1b301831&vid=c17e97ec30680cc9e622a04bff55a48e1b301831&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty